Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: BAZ1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013448.2 | NP_038476.2 | 24 | 4372 | ||
NM_182648.1 | NP_872589.1 | 23 | 4276 | |||
XM_011536374.1 | XP_011534676.1 | 23 | 3671 | |||
XM_011536376.2 | XP_011534678.2 | 14 | 2499 | |||
XM_017020942.1 | XP_016876431.1 | 13 | 2402 | |||
GenBank mRNA | AB032252.1 | - | 23 | 4273 | ||
AB209255.1 | - | 15 | 2623 | |||
AF213467.1 | - | 24 | 3970 | |||
AF221130.1 | - | 23 | 3804 | |||
AK027289.1 | - | 4 | 618 | |||
AL050089.1 | - | 7 | 1569 |