Search
Search

KLK8| Gene Symbol: KLK8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007196.3 | NP_009127.1 | 3 | 335 | ||
| NM_144505.2 | NP_653088.1 | 3 | 470 | |||
| NR_104008.1 | 3 | 335 | ||||
| GenBank mRNA | AB008390.2 | - | 3 | 210 | ||
| AB008927.1 | - | 2 | 312 | |||
| AB009849.1 | - | 3 | 190 | |||
| AF095742.1 | - | 3 | 335 | |||
| AY359036.1 | - | 3 | 321 | |||
| BC040887.1 | - | 4 | 646 | 5742016 | ||
| EU350538.1 | - | 3 | 237 | |||
| EU350539.1 | - | 3 | 372 | |||