Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Instrument Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
Our Instagram
Our Facebook
Blog Behind the Bench
Customer Experience Center (CEC)
Ecommerce Exclusives
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other LARP4 SIRNA Assays ›
Gene Symbol:
LARP4
Gene Name:
La ribonucleoprotein domain family member 4
Gene Aliases:
PP13296
Chromosome Location:
Chr.12: 50400809 - 50480005 on Build GRCh38
UniGene ID:
Hs.26613
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS174268
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Contact Us
Your Price
Online offer:
3062.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: LARP4
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001170803.1
NP_001164274.1
10
1146
NM_001170808.1
NP_001164279.1
11
1359
NM_052879.4
NP_443111.4
12
1572
NM_199188.2
NP_954658.2
12
1569
NM_199190.2
NP_954660.1
11
1359
XM_005268613.2
XP_005268670.1
13
1621
XM_011537828.2
XP_011536130.1
13
1676
XM_011537829.2
XP_011536131.1
13
1657
XM_011537830.2
XP_011536132.1
12
1504
XM_011537831.2
XP_011536133.1
12
1568
XM_011537833.2
XP_011536135.1
12
1463
XM_011537834.2
XP_011536136.1
12
1464
XM_011537835.2
XP_011536137.1
12
1444
XM_011537836.2
XP_011536138.1
12
1445
XM_011537838.2
XP_011536140.1
12
1416
XM_011537839.2
XP_011536141.1
13
1560
XM_011537840.2
XP_011536142.1
13
1613
XM_011537841.2
XP_011536143.1
12
1466
XM_011537843.2
XP_011536145.1
11
1232
XM_011537844.2
XP_011536146.1
13
1677
XM_017018750.1
XP_016874239.1
11
1285
XM_017018751.1
XP_016874240.1
13
1437
XM_017018752.1
XP_016874241.1
12
1420
XM_017018753.1
XP_016874242.1
12
1453
XM_017018754.1
XP_016874243.1
13
1603
XM_017018755.1
XP_016874244.1
11
1270
XM_017018756.1
XP_016874245.1
12
1322
XM_017018757.1
XP_016874246.1
13
1492
XM_017018758.1
XP_016874247.1
12
1428
XM_017018759.1
XP_016874248.1
11
1253
XM_017018760.1
XP_016874249.1
12
1572
XM_017018761.1
XP_016874250.1
11
1352
XM_017018762.1
XP_016874251.1
11
1352
GenBank mRNA
AF370416.1
-
12
1450
AK055521.1
-
10
1120
AK125113.1
-
11
1362
AK291746.1
-
12
1441
AL833272.1
-
12
1482
BC022377.1
-
10
1046
BX647457.1
-
11
1183
CR936626.1
-
12
1490
Back To Top
More Information
Panther Classification:
Molecular Function -
RNA metabolism protein
Gene Ontology Categories:
Function(s)
Process(es)
cytoskeleton organization
regulation of cell morphogenesis
nucleotide binding
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0