Search
Search

CLCA1| Gene Symbol: CLCA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001285.3 | NP_001276.2 | 7 | 1297 | ||
| GenBank mRNA | AF039400.1 | - | 8 | 1519 | ||
| AF127036.1 | - | 7 | 1172 | |||
| AK024970.1 | - | 2 | 381 | |||
| AK097057.1 | - | 7 | 1030 | |||
| AK300858.1 | - | 3 | 583 | |||
| AK314375.1 | - | 7 | 1215 | |||