Search
Search

CPXM2| Gene Symbol: CPXM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198148.2 | NP_937791.2 | 12, 13 | 2246 | ||
| XM_005269528.3 | XP_005269585.1 | 12, 13 | 2096 | |||
| XM_011539283.2 | XP_011537585.1 | 12, 13 | 2096 | |||
| XM_011539285.2 | XP_011537587.1 | 12, 13 | 1878 | |||
| XM_011539286.1 | XP_011537588.1 | 11, 12 | 1820 | |||
| XM_017015673.1 | XP_016871162.1 | 13, 14 | 1847 | |||
| XM_017015674.1 | XP_016871163.1 | 6, 7 | 1022 | |||
| GenBank mRNA | AK296365.1 | - | 12, 13 | 1653 | ||
| AK300506.1 | - | 8, 9 | 1207 | |||
| AK304023.1 | - | 13, 14 | 1945 | |||
| AK304814.1 | - | 12, 13 | 2057 | |||
| AL833732.1 | - | 10, 11 | 1424 | |||
| AY358565.1 | - | 12, 13 | 2246 | |||
| BC027623.1 | - | 14, 15 | 1969 | |||
| BC036789.1 | - | 12, 13 | 2076 | 5752707 | ||
| BC137492.1 | - | 12, 13 | 2241 | 9021492 | ||
| BC146862.1 | - | 12, 13 | 2241 | 9056790 | ||