Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: ZNF792 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175872.4 | NP_787068.3 | 4 | 2154 | ||
GenBank mRNA | AK095770.1 | - | 1 | 546 | ||
AK130901.1 | - | 1 | 863 | |||
BC101118.1 | - | 2 | 1550 | 40027074 | ||
BC101119.2 | - | 2 | 1550 | 40027075 | ||
BC101120.1 | - | 2 | 1550 | 40027081 |