Search
Search

CPA3| Gene Symbol: CPA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001870.3 | NP_001861.2 | 11 | 1183 | ||
| GenBank mRNA | AK313628.1 | - | 11 | 1143 | ||
| BC012613.1 | - | 11 | 1146 | 4104766 | ||
| BF508919.1 | - | 1 | 513 | |||
| BX105358.1 | - | 1 | 86 | |||
| M27717.1 | - | 11 | 1131 | |||
| S40234.1 | - | 8 | 804 | |||