Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: UBXN2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077619.1 | NP_001071087.1 | 8 | 971 | ||
XM_005251165.2 | XP_005251222.1 | 7 | 736 | |||
GenBank mRNA | AK126300.1 | - | 8 | 874 | ||
AL831990.2 | - | 8 | 845 | |||
AL833075.2 | - | 8 | 845 | |||
BC113645.1 | - | 8 | 971 | 8322697 | ||
BX647876.2 | - | 8 | 845 |
Gene Symbol: COL14A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | M64109.1 | - | 1296 |