Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Clinical Genomics
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CREM SIRNA Assays ›
Gene Symbol:
CREM
Gene Name:
cAMP responsive element modulator
Gene Aliases:
CREM-2, ICER, hCREM-2
Chromosome Location:
Chr.10: 35126841 - 35212958 on Build GRCh38
UniGene ID:
Hs.200250
Species:
Human
siRNA ID
HSS175184
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CREM
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001881.3
NP_001872.3
2, 3
475
NM_181571.2
NP_853549.1
3, 4
331
NM_183011.1
NP_898829.1
2, 3
220
NM_183012.1
NP_898830.1
2, 3
220
NM_183013.2
NP_898831.1
3, 4
573
NM_183060.2
NP_898883.1
2, 3
233
XM_006717378.3
XP_006717441.1
3, 4
313
XM_006717379.3
XP_006717442.1
3, 4
364
XM_006717382.3
XP_006717445.1
3, 4
305
XM_006717383.3
XP_006717446.1
3, 4
305
XM_006717387.3
XP_006717450.1
3, 4
305
XM_011519324.2
XP_011517626.1
3, 4
305
XM_011519325.2
XP_011517627.1
3, 4
387
XM_011519327.2
XP_011517629.1
2, 3
285
XM_017015716.1
XP_016871205.1
2, 3
424
XM_017015717.1
XP_016871206.1
2, 3
351
XM_017015718.1
XP_016871207.1
3, 4
305
XM_017015719.1
XP_016871208.1
2, 3
305
XM_017015720.1
XP_016871209.1
3, 4
305
XM_017015721.1
XP_016871210.1
2, 3
294
XM_017015722.1
XP_016871211.1
2, 3
207
XM_017015723.1
XP_016871212.1
2, 3
364
XM_017015726.1
XP_016871215.1
2, 3
232
GenBank mRNA
AV707433.1
-
2, 3
213
BC017117.1
-
2, 3
339
BC066342.1
-
2, 3
393
BG573680.1
-
2, 3
445
BI561543.1
-
333
D14825.1
-
3, 4
252
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
glucose metabolic process
transcription, DNA-templated
regulation of transcription, DNA-templated
fatty acid metabolic process
glycosphingolipid metabolic process
signal transduction
multicellular organism development
spermatogenesis
cell differentiation
circadian regulation of gene expression
regulation of circadian rhythm
negative regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
retinoic acid receptor signaling pathway
response to cAMP
core promoter sequence-specific DNA binding
DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding
cAMP response element binding protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-74fd55b88b-rxbn5:80/10.239.162.255:80.
git-commit: e4d250c089da5f210ba2ef291b70bc15861d50fd
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.9.0-2024.05.21-1.0