This product has been added to your favorites list. Go to
My Favorites
LOADING 
- Gene Symbol:
-
CSTF3
- Gene Name:
-
cleavage stimulation factor subunit 3
- Gene Aliases:
-
CSTF-77
- Chromosome Location:
-
Chr.11: 33084584 - 33161491 on Build GRCh38
- UniGene ID:
-
Hs.44402
- Species:
- Human
- Species Specific ID (Flybase ID):
-
-
siRNA Details
|
Gene Symbol:
CSTF3 |
| Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
| RefSeq |
NM_001033505.1 |
NP_001028677.1 |
1, 2
|
191
|
|
| NM_001033506.1 |
NP_001028678.1 |
1, 2
|
191
|
|
| NM_001326.2 |
NP_001317.1 |
1, 2
|
191
|
|
| GenBank mRNA |
AK290836.1 |
- |
1, 2
|
191
|
|
| AK297371.1 |
- |
1, 2
|
186
|
|
| BC009792.1 |
- |
1, 2
|
173
|
|
| BC059948.1 |
- |
1, 2
|
45
|
|
| BC108319.1 |
- |
1, 2
|
165
|
6197136 |
| BG435786.1 |
- |
1, 2
|
192
|
|
| U15782.1 |
- |
1, 2
|
156
|
|
Back To Top
Related Products
-
Anti-GAPDH, Mouse Monoclonal 6C5
-
Anti-β-Actin, Mouse Monoclonal AC-15
-
TaqMan® Gene Expression Cells-to-CT™
-
Neon™ Transfection System 100 µL Kit
-
Lipofectamine™ RNAiMAX Transfection Reagent
-
Invivofectamine 2.0
-
Lipofectamine™ 2000 Transfection Reagent
-
Neon™ Transfection System