Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other MEI1 SIRNA Assays ›
          Gene Symbol:
          MEI1
          Gene Name:
          meiotic double-stranded break formation protein 1
          Gene Aliases:
          SPATA38
          Chromosome Location:
          Chr.22: 41699503 - 41799456 on Build GRCh38
          UniGene ID:
          Hs.116419
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS175625
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: MEI1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_152513.3 NP_689726.3 6, 7 747
          XM_011529935.1 XP_011528237.1 6, 7 756
          XM_011529936.1 XP_011528238.1 6, 7 756
          XM_011529937.1 XP_011528239.1 6, 7 756
          XM_011529938.1 XP_011528240.1 6, 7 756
          XM_011529939.1 XP_011528241.1 6, 7 756
          XM_011529940.1 XP_011528242.1 6, 7 756
          XM_011529941.1 XP_011528243.1 6, 7 756
          XM_011529942.2 XP_011528244.1 6, 7 947
          XM_011529943.1 XP_011528245.1 6, 7 756
          XM_011529944.1 XP_011528246.1 6, 7 756
          XM_011529945.2 XP_011528247.1 6, 7 758
          XM_011529946.1 XP_011528248.1 6, 7 756
          XM_011529947.1 XP_011528249.1 6, 7 756
          XM_011529948.2 XP_011528250.1 2, 3 128
          XM_011529950.1 XP_011528252.1 6, 7 756
          XM_011529952.1 XP_011528254.1 1, 2 195
          XM_011529955.1 XP_011528257.1 6, 7 756
          XM_011529956.2 XP_011528258.1 6, 7 756
          XM_017028633.1 XP_016884122.1 6, 7 756
          GenBank mRNA BC025400.1 - 6, 7 763
          BC035720.1 - 6, 7 746
          BC070111.1 - 1, 2 197

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s)

          meiosis I
          male meiosis I
          spermatid development
          meiotic telomere clustering

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-269v7:80/100.66.79.229:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0