Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other USP54 SIRNA Assays ›
          Gene Symbol:
          USP54
          Gene Name:
          ubiquitin specific peptidase 54
          Gene Aliases:
          C10orf29, bA137L10.3, bA137L10.4
          Chromosome Location:
          Chr.10: 73497538 - 73626844 on Build GRCh38
          UniGene ID:
          Hs.657355
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS175894
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: USP54
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001320437.1 NP_001307366.1 12 1387
          NM_001320441.1 NP_001307370.1 14 2122
          NM_152586.3 NP_689799.3 12 1485
          NR_135249.1 15 2188
          NR_135250.1 10 1183
          XM_005269582.2 XP_005269639.1 11 4563
          XM_011539368.2 XP_011537670.1 12 4629
          XM_017015759.1 XP_016871248.1 14 4058
          XM_017015760.1 XP_016871249.1 14 2960
          XM_017015761.1 XP_016871250.1 14 3798
          XM_017015762.1 XP_016871251.1 14 2968
          XM_017015763.1 XP_016871252.1 14 2262
          XM_017015764.1 XP_016871253.1 14 2325
          XM_017015765.1 XP_016871254.1 15 2578
          XM_017015766.1 XP_016871255.1 14 4184
          XM_017015767.1 XP_016871256.1 14 3843
          XM_017015768.1 XP_016871257.1 14 2014
          XM_017015769.1 XP_016871258.1 14 1622
          XM_017015770.1 XP_016871259.1 14 2283
          XM_017015771.1 XP_016871260.1 13 4800
          XM_017015772.1 XP_016871261.1 13 4806
          XM_017015773.1 XP_016871262.1 12 4740
          XM_017015774.1 XP_016871263.1 13 3726
          XM_017015775.1 XP_016871264.1 14 3604
          XM_017015776.1 XP_016871265.1 13 4109
          XM_017015777.1 XP_016871266.1 13 3768
          XM_017015778.1 XP_016871267.1 13 4806
          XM_017015779.1 XP_016871268.1 13 4806
          XM_017015780.1 XP_016871269.1 13 4806
          XM_017015781.1 XP_016871270.1 12 4635
          XM_017015782.1 XP_016871271.1 14 2506
          XM_017015783.1 XP_016871272.1 13 3433
          XM_017015784.1 XP_016871273.1 12 1379
          XM_017015785.1 XP_016871274.1 13 4806
          XM_017015786.1 XP_016871275.1 13 1996
          GenBank mRNA AK126497.1 - 2 156
          AK127267.1 - 10 1183
          AK302598.1 - 14 2312
          BX649087.1 - 13 2029

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          cysteine protease

          Gene Ontology Categories:

          Function(s) Process(es)

          protein deubiquitination
          protein binding
          thiol-dependent ubiquitinyl hydrolase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-9c8cf:80/100.66.75.150:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0