Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other DAG1 SIRNA Assays ›
          Gene Symbol:
          DAG1
          Gene Name:
          dystroglycan 1
          Gene Aliases:
          156DAG, A3a, AGRNR, DAG, MDDGA9, MDDGC7, MDDGC9
          Chromosome Location:
          Chr.3: 49468703 - 49535618 on Build GRCh38
          UniGene ID:
          Hs.76111
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS175913
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price Contact Us
          Your Price
          Online offer:
          3062.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: DAG1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001165928.3 NP_001159400.2 6 2655
          NM_001177634.2 NP_001171105.1 6 2598
          NM_001177635.2 NP_001171106.1 4 2187
          NM_001177636.2 NP_001171107.1 5 2576
          NM_001177637.2 NP_001171108.1 4 2414
          NM_001177638.2 NP_001171109.1 4 2445
          NM_001177639.2 NP_001171110.1 3 2135
          NM_001177640.2 NP_001171111.1 4 2351
          NM_001177641.2 NP_001171112.1 4 2441
          NM_001177642.2 NP_001171113.1 4 2346
          NM_001177643.2 NP_001171114.1 3 2345
          NM_001177644.2 NP_001171115.1 3 2434
          NM_004393.5 NP_004384.4 3 2294
          GenBank mRNA AK291692.1 - 3 2136
          BC012740.2 - 3 2229 4099382
          BC014616.2 - 3 2229 2964740
          L19711.1 - 3 2270

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          cell adhesion molecule

          Gene Ontology Categories:

          Function(s) Process(es)

          morphogenesis of an epithelial sheet
          protein O-linked glycosylation
          membrane protein ectodomain proteolysis
          NLS-bearing protein import into nucleus
          cytoskeletal anchoring at plasma membrane
          regulation of gastrulation
          regulation of epithelial to mesenchymal transition
          calcium-dependent cell-matrix adhesion
          regulation of embryonic cell shape
          modulation by virus of host morphology or physiology
          nerve maturation
          myelination in peripheral nervous system
          extracellular matrix organization
          negative regulation of cell migration
          microtubule anchoring
          negative regulation of MAPK cascade
          response to peptide hormone
          viral entry into host cell
          negative regulation of protein kinase B signaling
          epithelial tube branching involved in lung morphogenesis
          branching involved in salivary gland morphogenesis
          commissural neuron axon guidance
          basement membrane organization
          positive regulation of basement membrane assembly involved in embryonic body morphogenesis
          virus receptor activity
          dystroglycan binding
          actin binding
          calcium ion binding
          protein binding
          structural constituent of muscle
          tubulin binding
          vinculin binding
          protein complex binding
          SH2 domain binding
          laminin-1 binding
          alpha-actinin binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-fz5f2:80/100.66.72.225:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0