Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: UGT3A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168316.1 | NP_001161788.1 | 2, 3 | 332 | ||
NM_174914.3 | NP_777574.2 | 3, 4 | 434 | |||
XM_011513988.2 | XP_011512290.1 | 4, 5 | 476 | |||
XM_017009150.1 | XP_016864639.1 | 3, 4 | 485 | |||
GenBank mRNA | AK075383.1 | - | 3, 4 | 395 | ||
AK300751.1 | - | 2, 3 | 293 | |||
AY358416.1 | - | 3, 4 | 369 | |||
BC103924.1 | - | 3, 4 | 383 | 40007344 | ||
BC103925.1 | - | 3, 4 | 383 | 40007348 | ||
DB110369.1 | - | 3, 4 | 435 |