Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other EEF1D SIRNA Assays ›
          Gene Symbol:
          EEF1D ANKRD19P
          Gene Name:
          eukaryotic translation elongation factor 1 delta
          ankyrin repeat domain 19, pseudogene
          Gene Aliases:
          EF-1D, EF1D, FP1047
          ANKRD19, bA526D8.2
          Chromosome Location:
          Chr.8: 143579697 - 143597675 on Build GRCh38
          Chromosome Location:
          Chr.9: 92809611 - 92838457 on Build GRCh38
          UniGene ID:
          Hs.333388
          Hs.643597
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID HSS176327
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price Contact Us
          Your Price
          Online offer:
          3062.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: EEF1D
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001130053.2 NP_001123525.2 4 1515
          NM_001130055.2 NP_001123527.1 3 586
          NM_001130056.2 NP_001123528.1 2 410
          NM_001130057.2 NP_001123529.1 2 592
          NM_001195203.1 NP_001182132.1 2 410
          NM_001289950.1 NP_001276879.1 2 407
          NM_001317743.1 NP_001304672.1 2 407
          NM_001960.4 NP_001951.2 2 410
          NM_032378.4 NP_115754.3 4 1632
          XM_005250820.3 XP_005250877.1 3 6507
          XM_005250823.1 XP_005250880.1 3 1241
          XM_006716519.3 XP_006716582.1 3 6504
          XM_006716520.3 XP_006716583.1 3 6507
          XM_006716522.1 XP_006716585.1 3 1495
          XM_006716523.1 XP_006716586.1 4 1455
          XM_006716524.1 XP_006716587.1 3 1262
          XM_006716525.1 XP_006716588.1 2 430
          XM_011516905.2 XP_011515207.1 4 6077
          XM_011516906.2 XP_011515208.1 6 4669
          XM_017013168.1 XP_016868657.1 4 5508
          XM_017013169.1 XP_016868658.1 5 5164
          XM_017013170.1 XP_016868659.1 4 4950
          XM_017013171.1 XP_016868660.1 3 1238
          XM_017013172.1 XP_016868661.1 3 1495
          XM_017013173.1 XP_016868662.1 2 587
          GenBank mRNA AF370363.1 - 4 1215
          AK024550.1 - 4 1369
          AK093756.1 - 2 1552
          AK225040.1 - 4 1370
          AK225243.1 - 2 147
          AK225270.1 - 2 148
          AK293339.1 - 2 148
          AK314311.1 - 4 1251
          BC000678.2 - 2 954
          BC007847.2 - 4 1236 4129368
          BC009907.2 - 1 132
          BC012819.2 - 1 132
          BC062535.1 - 2 124
          BC071840.1 - 2 112
          BC094806.1 - 2 121
          BG482145.1 - 2 114
          BG748894.1 - 2 592
          BI667117.1 - 2 415
          BQ064320.1 - 267
          BQ686171.1 - 3 305
          BT007242.1 - 2 1173
          CF128478.1 - 2 497
          CN424074.1 - 2 399
          CN424083.1 - 2 351
          CN424105.1 - 2 195
          CX787126.1 - 2 457
          Z21507.1 - 2 144
          Gene Symbol: ANKRD19P
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_026868.1 9 1956
          GenBank mRNA AK093497.1 - 7 1548
          BC028712.1 - 9 1920
          BC038951.1 - 9 1367 5733799

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_001013104, XM_008765551, XM_008765557, XM_008765549, XM_008765554, and others.

          Panther Classification:

          Molecular Function -

          translation elongation factor

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          translational elongation
          signal transduction
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          cellular response to ionizing radiation
          cell-cell adhesion
          DNA binding
          translation elongation factor activity
          signal transducer activity
          protein binding
          translation factor activity, RNA binding
          cadherin binding involved in cell-cell adhesion

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0