Search
Search

F8| Gene Symbol: F8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000132.3 | NP_000123.1 | 25 | 6960 | ||
| NM_019863.2 | NP_063916.1 | 4 | 529 | |||
| GenBank mRNA | AK289947.1 | - | 4 | 459 | ||
| AK313707.1 | - | 4 | 414 | |||
| AX746360.1 | - | 25 | 6789 | |||
| BC022513.1 | - | 4 | 448 | |||
| BC064380.1 | - | 4 | 508 | |||
| BC098389.1 | - | 4 | 527 | |||
| BC111967.1 | - | 4 | 417 | |||
| BC111969.1 | - | 4 | 416 | |||
| K01740.1 | - | 25 | 6939 | |||
| KT878539.1 | - | 2 | 210 | |||
| M14113.1 | - | 25 | 6960 | |||
| M90707.1 | - | 4 | 424 | |||
| X01179.1 | - | 25 | 6898 | |||