Search
Search

FCGR2A| Gene Symbol: FCGR2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001136219.1 | NP_001129691.1 | 1 | 104 | ||
| NM_021642.3 | NP_067674.2 | 1 | 104 | |||
| XM_011509287.2 | XP_011507589.1 | 1 | 89 | |||
| XM_011509290.2 | XP_011507592.1 | 1 | 89 | |||
| XM_011509291.1 | XP_011507593.1 | 1 | 89 | |||
| XM_017000663.1 | XP_016856152.1 | 1 | 89 | |||
| XM_017000664.1 | XP_016856153.1 | 1 | 89 | |||
| XM_017000665.1 | XP_016856154.1 | 1 | 89 | |||
| XM_017000666.1 | XP_016856155.1 | 1 | 89 | |||
| XM_017000667.1 | XP_016856156.1 | 1 | 89 | |||
| XM_017000668.1 | XP_016856157.1 | 1 | 89 | |||
| GenBank mRNA | AK225438.1 | - | 1 | 62 | ||
| AK225601.1 | - | 1 | 60 | |||
| AK226059.1 | - | 1 | 76 | |||
| AK298657.1 | - | 1 | 62 | |||
| AK298734.1 | - | 1 | 54 | |||
| AK315225.1 | - | 1 | 62 | |||
| BC019931.1 | - | 1 | 89 | 4536764 | ||
| BC020823.1 | - | 1 | 54 | 4704252 | ||
| J03619.1 | - | 1 | 56 | |||
| M28697.1 | - | 1 | 38 | |||
| M31932.1 | - | 1 | 58 | |||
| X62572.1 | - | 1 | 38 | |||
| Y00644.1 | - | 1 | 70 | |||