Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ?
立即註冊
產品
Antibodies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Cell Culture Media
Chemicals
Western Blot Products
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Greener Products
查看所有產品類別
應用
生物製程
細胞培養和轉染
食品和飲料
實驗室解決方案
製藥和生物製藥
即時定量PCR
半導體分析
臨床和診斷
數位解決方案
分子光譜、同位素與元素分析
查看所有應用與技術
服務
360° CDMO 和 CRO 服務
CDMO 服务
临床试验 CRO 服务
客製化服務
企業服務
金融與租賃服務
儀器服務
實驗室資訊學
授權與商業供應
培訓服務
查看所有服務
支援
線上訂購協助
數位解決方案
產品支持
技術相關資訊
培訓與教育
查看所有協助與支援
熱門商品
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
特價商品
聯絡我們
快速訂購
訂單狀態與貨物追蹤
文件和證書
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
訂單查詢
快速訂購
登入
登入
還沒有帳戶嗎 ?
立即註冊
帳戶
訂單
Connect: Lab, Data, Apps
客製化產品與項目
儀器管理
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other FOXJ1 SIRNA Assays ›
Gene Symbol:
FOXJ1
Gene Name:
forkhead box J1
Gene Aliases:
FKHL13, HFH-4, HFH4
Chromosome Location:
Chr.17: 76136333 - 76141299 on Build GRCh38
UniGene ID:
Hs.651204
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS177105
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
新增到購物車
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: FOXJ1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001454.3
NP_001445.2
3
1013
GenBank mRNA
BC046460.1
-
3
1004
5171637
U69537.1
-
2
748
X99349.1
-
2
666
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
central tolerance induction
negative regulation of germinal center formation
positive regulation of central B cell tolerance induction
negative regulation of humoral immune response mediated by circulating immunoglobulin
transcription from RNA polymerase II promoter
humoral immune response
spermatogenesis
determination of left/right symmetry
pattern specification process
brain development
heart development
actin cytoskeleton organization
negative regulation of NF-kappaB transcription factor activity
negative regulation of T cell differentiation in thymus
establishment of apical/basal cell polarity
metanephric part of ureteric bud development
negative regulation of T cell proliferation
cilium assembly
negative regulation of interleukin-6 biosynthetic process
positive regulation of transcription from RNA polymerase II promoter
negative regulation of B cell activation
leukocyte migration
cilium morphogenesis
lung epithelium development
epithelium development
left/right pattern formation
glomerular parietal epithelial cell development
activation of GTPase activity
positive regulation of lung ciliated cell differentiation
transcription regulatory region sequence-specific DNA binding
RNA polymerase II core promoter proximal region sequence-specific DNA binding
RNA polymerase II transcription factor activity, sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
DNA binding
transcription factor activity, sequence-specific DNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-sws2h:80/100.66.79.160:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0