Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: SSPO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198455.2 | NP_940857.2 | 101 | 15218 | ||
GenBank mRNA | AK093431.1 | - | 10 | 2167 | ||
AK123170.1 | - | 17 | 2233 | |||
AK124542.1 | - | 8 | 3103 |