Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CYFIP1 SIRNA Assays ›
          Gene Symbol:
          CYFIP1
          Gene Name:
          cytoplasmic FMR1 interacting protein 1
          Gene Aliases:
          P140SRA-1, SHYC, SRA-1, SRA1
          Chromosome Location:
          Chr.15: 22869465 - 22980906 on Build GRCh38
          UniGene ID:
          Hs.26704
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS177242
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price Contact Us
          Your Price
          Online offer:
          3062.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CYFIP1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001033028.1 NP_001028200.1 11 2027
          NM_001287810.2 NP_001274739.1 27 3298
          NM_001324119.1 NP_001311048.1 26 3127
          NM_001324120.1 NP_001311049.1 26 3306
          NM_001324122.1 NP_001311051.1 27 3327
          NM_001324123.1 NP_001311052.1 26 3206
          NM_001324124.1 NP_001311053.1 25 3089
          NM_001324125.1 NP_001311054.1 23 2888
          NM_001324126.1 NP_001311055.1 25 3077
          NM_014608.4 NP_055423.1 26 3179
          XM_011543873.2 XP_011542175.1 26 3528
          XM_011543874.1 XP_011542176.1 26 3561
          XM_011543876.2 XP_011542178.2 26 3129
          XM_017022023.1 XP_016877512.1 26 3534
          XM_017022024.1 XP_016877513.1 26 3542
          GenBank mRNA AK125784.1 - 5 2165
          AK127094.1 - 11 2027
          AK291604.1 - 26 3106
          AY763577.1 - 26 3143
          AY763578.1 - 27 3209
          AY763579.1 - 11 2027
          AY763580.1 - 5 2165
          BC001306.2 - 9 995
          BC005097.1 - 26 3070 3163591
          BC063869.1 - 11 1821
          D38549.1 - 26 3073
          KJ534819.1 - 25 3018

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          scaffold/adaptor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of cell shape
          Rac protein signal transduction
          lamellipodium assembly
          ruffle organization
          Fc-gamma receptor signaling pathway involved in phagocytosis
          vascular endothelial growth factor receptor signaling pathway
          axon extension
          cognition
          positive regulation of Arp2/3 complex-mediated actin nucleation
          protein binding
          protein complex binding
          Rac GTPase binding
          actin filament binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0