Search Thermo Fisher Scientific
Gene Symbol: RRP12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015179.3 | NP_055994.2 | 5 | 714 | ||
XM_011539555.1 | XP_011537857.1 | 4 | 583 | |||
XM_011539556.2 | XP_011537858.1 | 5 | 684 | |||
GenBank mRNA | AK000238.1 | - | 2 | 124 | ||
AK131283.1 | - | 1 | 54 | |||
BC012745.2 | - | 5 | 652 | 4025242 |