Search
Search

AHCYL2| Gene Symbol: AHCYL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001130720.2 | NP_001124192.1 | 16 | 1792 | ||
| NM_001130722.2 | NP_001124194.2 | 16 | 1650 | |||
| NM_001130723.2 | NP_001124195.1 | 16 | 1795 | |||
| NM_015328.3 | NP_056143.1 | 16 | 1795 | |||
| XM_005250237.1 | XP_005250294.1 | 16 | 1798 | |||
| XM_011515987.2 | XP_011514289.1 | 17 | 2769 | |||
| XM_011515988.2 | XP_011514290.1 | 17 | 2736 | |||
| XM_017011904.1 | XP_016867393.1 | 16 | 1513 | |||
| XM_017011905.1 | XP_016867394.1 | 16 | 1673 | |||
| XM_017011906.1 | XP_016867395.1 | 17 | 1689 | |||
| XM_017011907.1 | XP_016867396.1 | 17 | 1497 | |||
| GenBank mRNA | AB020635.1 | - | 16 | 1756 | ||
| AK025372.1 | - | 16 | 1694 | |||
| AK295851.1 | - | 16 | 1484 | |||
| AK316073.1 | - | 16 | 1650 | |||
| BC008349.1 | - | 16 | 1734 | 3536052 | ||
| BC024325.1 | - | 16 | 1786 | 3907552 | ||