Search
Search

EDC4| Gene Symbol: EDC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014329.4 | NP_055144.3 | 7 | 1084 | ||
| GenBank mRNA | AK128582.1 | - | 7 | 1238 | ||
| AK291049.1 | - | 7 | 852 | |||
| AK302563.1 | - | 7 | 850 | |||
| BC043616.1 | - | 7 | 987 | 6065418 | ||
| BC053598.1 | - | 2 | 224 | |||
| BC064567.1 | - | 7 | 981 | 5575195 | ||
| L26339.1 | - | 7 | 981 | |||