Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Pipettes and Pipette Tips
Lab Centrifuges
Ultra-Low Temperature Freezers
Spectroscopy
Beakers
PCR Equipment and Supplies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all product categories
Applications
Cell Analysis
Lab Equipment
Real-Time PCR
PCR
Chromatography
Cell Culture and Transfection
DNA and RNA Extraction and Analysis
Protein Biology
Flow Cytometry
Chemicals
See all applications and techniques
Services
Custom Services
Lab Informatics
Enterprise Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Instrument Services
Training Services
See all services
Help and Support
Customer Center
Contact Us
Certificates of Analysis and Conformance
Safety Data Sheets (SDS)
Manuals
How to Cite Our Products in a Paper
Instrument Support
Knowledge Base and Product FAQs
Learning Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RALGAPA1 SIRNA Assays ›
Gene Symbol:
RALGAPA1
Gene Name:
Ral GTPase activating protein catalytic alpha subunit 1
Gene Aliases:
GARNL1, GRIPE, RalGAPalpha1, TULIP1, p240
Chromosome Location:
Chr.14: 35538352 - 35809304 on Build GRCh38
UniGene ID:
Hs.113150
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS177699
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: RALGAPA1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001283043.1
NP_001269972.1
1, 2
500
NM_001283044.1
NP_001269973.1
1, 2
500
NM_014990.1
NP_055805.1
1, 2
500
NM_194301.2
NP_919277.2
1, 2
500
XM_005267491.3
XP_005267548.1
1, 2
578
XM_005267492.3
XP_005267549.1
1, 2
578
XM_005267493.3
XP_005267550.1
1, 2
578
XM_006720098.2
XP_006720161.1
1, 2
578
XM_006720099.2
XP_006720162.1
1, 2
578
XM_006720100.3
XP_006720163.1
1, 2
578
XM_006720101.2
XP_006720164.1
1, 2
578
XM_006720102.3
XP_006720165.1
1, 2
284
XM_006720103.2
XP_006720166.1
1, 2
578
XM_006720104.3
XP_006720167.1
1, 2
578
XM_011536615.2
XP_011534917.1
1, 2
578
XM_011536616.2
XP_011534918.1
1, 2
578
XM_017021141.1
XP_016876630.1
1, 2
578
XM_017021142.1
XP_016876631.1
1, 2
578
XM_017021143.1
XP_016876632.1
1, 2
578
XM_017021144.1
XP_016876633.1
1, 2
578
XM_017021145.1
XP_016876634.1
1, 2
578
GenBank mRNA
AB511280.1
-
1, 2
109
AY596970.1
-
1, 2
500
AY596971.1
-
1, 2
500
BC150596.1
-
1, 2
208
9056966
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_001003719, NM_001112714, NM_001286263, NM_019994, BC145119, and others.
Panther Classification:
Molecular Function -
GTPase-activating protein
Gene Ontology Categories:
Function(s)
Process(es)
regulation of transcription, DNA-templated
regulation of small GTPase mediated signal transduction
activation of GTPase activity
GTPase activator activity
protein heterodimerization activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-hdb68:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0