Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture and Transfection
Chemicals
Chromatography
Electron Microscopes
Lab Plasticware and Supplies
Lab Centrifuges
Lab Solutions
Mass Spectrometers
Next Generation Sequencers
See all product categories
Applications
Brands
Industrial and Applied Sciences
Food and Beverage
Forensics
Lab Solutions
Life Sciences
Pharma and Biopharma
Biotechnology
Clinical and Diagnostics
Digital Solutions
See all applications
Services
Lab Instrument and Equipment Services
Custom Services
Training Services
Financial and Leasing Services
Enterprise Level Lab Informatics
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Cell Biology Services
Food Safety Inspection Services
See all services
Help and Support
Contact Us
Product Documentation
Knowledge Base and Product FAQs
Learning Centers
Supply Center
eProcurement Solutions
Lab Instrument and Equipment Support
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other LETMD1 SIRNA Assays ›
Gene Symbol:
LETMD1
Gene Name:
LETM1 domain containing 1
Gene Aliases:
1110019O13Rik, HCCR-1, HCCR-2, HCCR1, HCRR-2
Chromosome Location:
Chr.12: 51048230 - 51068984 on Build GRCh38
UniGene ID:
Hs.655272
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS177884
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Contact Us
Your Price
Online offer:
3062.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: LETMD1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001243689.1
NP_001230618.1
7
994
NM_001300765.1
NP_001287694.1
4
583
NM_015416.4
NP_056231.3
7
955
NR_045017.1
7
830
NR_045018.1
6
839
NR_045019.1
5
756
NR_045020.1
3
467
XM_006719335.1
XP_006719398.1
6
777
XM_006719336.1
XP_006719399.1
6
665
XM_006719339.1
XP_006719402.1
7
893
XM_006719340.1
XP_006719403.1
6
714
XM_011538161.2
XP_011536463.1
7
962
XM_011538162.2
XP_011536464.1
7
963
XM_011538163.2
XP_011536465.1
7
964
XM_011538164.2
XP_011536466.1
6
812
XM_011538165.2
XP_011536467.1
4
590
XM_011538167.1
XP_011536469.1
2
384
XM_017019151.1
XP_016874640.1
7
964
XM_017019152.1
XP_016874641.1
7
930
XM_017019153.1
XP_016874642.1
6
814
XM_017019154.1
XP_016874643.1
6
777
XM_017019155.1
XP_016874644.1
7
802
XM_017019156.1
XP_016874645.1
5
662
XM_017019157.1
XP_016874646.1
5
624
XM_017019158.1
XP_016874647.1
6
665
XM_017019159.1
XP_016874648.1
4
593
XM_017019160.1
XP_016874649.1
4
555
XM_017019161.1
XP_016874650.1
4
561
XM_017019162.1
XP_016874651.1
4
531
XM_017019163.1
XP_016874652.1
5
624
XM_017019164.1
XP_016874653.1
6
831
XM_017019165.1
XP_016874654.1
7
893
XM_017019166.1
XP_016874655.1
6
714
XM_017019167.1
XP_016874656.1
4
555
XM_017019168.1
XP_016874657.1
4
555
GenBank mRNA
AF195651.2
-
7
927
AF315598.1
-
7
791
AF329277.1
-
6
838
AK000639.1
-
7
942
AK057866.1
-
7
900
AK127540.1
-
7
955
AK225836.1
-
7
913
AK301955.1
-
7
830
AK304707.1
-
3
439
AK315119.1
-
7
922
AL050286.1
-
2
447
AY259835.1
-
5
698
AY259836.1
-
3
409
BC000395.2
-
2
204
BC019274.1
-
2
204
BC064943.1
-
7
901
6009854
BM549678.1
-
439
BX343550.2
-
5
697
BX376670.2
-
4
524
Back To Top
More Information
Panther Classification:
Molecular Function -
secondary carrier transporter
Gene Ontology Categories:
Process(es)
protein binding
ribosome binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0