Search Thermo Fisher Scientific
Gene Symbol: LSM14A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114093.1 | NP_001107565.1 | 7 | 1129 | ||
NM_015578.2 | NP_056393.2 | 7 | 1129 | |||
XM_005258719.4 | XP_005258776.1 | 7 | 1148 | |||
XM_005258720.4 | XP_005258777.1 | 6 | 971 | |||
XM_017026575.1 | XP_016882064.1 | 6 | 971 | |||
XM_017026576.1 | XP_016882065.1 | 6 | 971 | |||
GenBank mRNA | AB069974.1 | - | 7 | 1008 | ||
AB069975.1 | - | 7 | 1008 | |||
AK027369.1 | - | 6 | 739 | |||
AK027643.1 | - | 7 | 1010 | |||
AK127401.1 | - | 4 | 1252 | |||
AK300208.1 | - | 6 | 928 | |||
AL117499.1 | - | 1 | 13 | |||
AL833221.1 | - | 5 | 602 | |||
AL834398.1 | - | 7 | 1038 | |||
BC016842.1 | - | 7 | 1129 | 3902704 |