Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: RIBC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015653.4 | NP_056468.3 | 5 | 930 | ||
XM_005261524.4 | XP_005261581.1 | 5 | 755 | |||
XM_011530126.2 | XP_011528428.1 | 3 | 458 | |||
GenBank mRNA | AK098586.1 | - | 5 | 930 | ||
AL442116.1 | - | 5 | 881 | |||
BC003024.1 | - | 5 | 892 | 2822570 | ||
BC009904.2 | - | 5 | 892 | 2822570 | ||
CR457398.1 | - | 3 | 515 |