Search
Search

GCG| Gene Symbol: GCG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002054.4 | NP_002045.1 | 2 | 314 | ||
| GenBank mRNA | BC005278.1 | - | 2 | 157 | 3950435 | |
| BM272442.1 | - | 2 | 317 | |||
| BT006813.1 | - | 1 | 58 | |||
| J04040.1 | - | 2 | 95 | |||
| X05388.1 | - | 1 | 67 | |||