Search
Search

PDLIM3| Gene Symbol: PDLIM3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001114107.4 | NP_001107579.1 | 6 | 855 | ||
| NM_001257962.1 | NP_001244891.1 | 6 | 735 | |||
| NM_001257963.1 | NP_001244892.1 | 4 | 498 | |||
| NM_014476.5 | NP_055291.2 | 7 | 999 | |||
| NR_047562.1 | 5 | 667 | ||||
| GenBank mRNA | AF002280.1 | - | 7 | 921 | ||
| AF002282.1 | - | 6 | 777 | |||
| AF039018.1 | - | 7 | 945 | |||
| AK129863.1 | - | 5 | 536 | |||
| AK222966.1 | - | 7 | 903 | |||
| AK313253.1 | - | 7 | 948 | |||
| BC027870.1 | - | 6 | 782 | 5205215 | ||
| BX647263.1 | - | 3 | 4376 | |||
| CD625728.1 | - | 2 | 202 | |||
| DA898089.1 | - | 4 | 447 | |||