Search
Search

FADS6| Gene Symbol: FADS6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178128.5 | NP_835229.3 | 4, 5 | 1004 | ||
| XM_005257224.4 | XP_005257281.2 | 4, 5 | 998 | |||
| XM_017024458.1 | XP_016879947.1 | 5, 6 | 779 | |||
| GenBank mRNA | AK094411.1 | - | 4, 5 | 852 | ||
| AY203929.1 | - | 4, 5 | 813 | |||
| BC117230.1 | - | 4, 5 | 833 | 40125781 | ||