Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: SLC26A11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166347.1 | NP_001159819.1 | 9, 10 | 1294 | ||
NM_001166348.1 | NP_001159820.1 | 8, 9 | 1224 | |||
NM_001166349.1 | NP_001159821.1 | 8, 9 | 1094 | |||
NM_173626.3 | NP_775897.3 | 9, 10 | 1291 | |||
XM_006721833.1 | XP_006721896.1 | 9, 10 | 1366 | |||
XM_011524652.1 | XP_011522954.1 | 7, 8 | 1127 | |||
XM_011524653.1 | XP_011522955.1 | 7, 8 | 1055 | |||
XM_011524654.1 | XP_011522956.1 | 7, 8 | 940 | |||
XM_017024505.1 | XP_016879994.1 | 8, 9 | 1280 | |||
XM_017024506.1 | XP_016879995.1 | 8, 9 | 1280 | |||
XM_017024507.1 | XP_016879996.1 | 9, 10 | 1340 | |||
GenBank mRNA | AF331524.1 | - | 1, 2 | 68 | ||
AK075248.1 | - | 9, 10 | 1254 | |||
AK315132.1 | - | 9, 10 | 1257 | |||
AL833468.1 | - | 8, 9 | 1188 | |||
BC035900.1 | - | 9, 10 | 1233 | 5241643 | ||
BC047451.1 | - | 3, 4 | 343 |