Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: C8orf31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173687.3 | NP_775958.1 | 2 | 444 | ||
NR_126398.1 | 2 | 444 | ||||
XM_017013344.1 | XP_016868833.1 | 2 | 670 | |||
XM_017013345.1 | XP_016868834.1 | 1 | 65 | |||
XM_017013346.1 | XP_016868835.1 | 1 | 65 | |||
GenBank mRNA | AK094450.1 | - | 2 | 392 | ||
BC073830.1 | - | 2 | 392 |