Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Pipettes and Pipette Tips
Lab Centrifuges
Ultra-Low Temperature Freezers
Spectroscopy
Beakers
PCR Equipment and Supplies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all product categories
Applications
Cell Analysis
Lab Equipment
Real-Time PCR
PCR
Chromatography
Cell Culture and Transfection
DNA and RNA Extraction and Analysis
Protein Biology
Flow Cytometry
Chemicals
See all applications and techniques
Services
Custom Services
Lab Informatics
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Instrument Services
Training Services
Unity Lab Services
See all services
Help and Support
Customer Center
Contact Us
Certificates of Analysis and Conformance
Safety Data Sheets (SDS)
Manuals
How to Cite Our Products in a Paper
Instrument Support
Knowledge Base and Product FAQs
Learning Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ANK1 SIRNA Assays ›
Gene Symbol:
ANK1
Gene Name:
ankyrin 1
Gene Aliases:
ANK, SPH1, SPH2
Chromosome Location:
Chr.8: 41653225 - 41896762 on Build GRCh38
UniGene ID:
Hs.654438
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS178894
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ANK1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_000037.3
NP_000028.3
40
5514
NM_001142445.1
NP_001135917.1
2
518
NM_001142446.1
NP_001135918.1
41
5835
NM_020475.2
NP_065208.2
40
5514
NM_020476.2
NP_065209.2
40
5514
NM_020477.2
NP_065210.2
40
5028
NM_020478.4
NP_065211.2
2
518
NM_020480.4
NP_065213.2
2
518
XM_005273476.4
XP_005273533.1
41
5812
XM_011544490.2
XP_011542792.1
43
5779
XM_011544491.2
XP_011542793.1
43
5778
XM_011544494.2
XP_011542796.1
42
5733
XM_011544495.2
XP_011542797.1
42
5732
XM_011544496.2
XP_011542798.1
43
5779
XM_011544500.2
XP_011542802.1
40
5616
XM_011544501.2
XP_011542803.1
40
5616
XM_011544502.2
XP_011542804.1
40
5616
XM_011544503.2
XP_011542805.1
42
5245
XM_011544504.2
XP_011542806.1
40
5130
XM_011544505.2
XP_011542807.1
40
5130
XM_017013319.1
XP_016868808.1
42
5754
XM_017013320.1
XP_016868809.1
43
5780
XM_017013321.1
XP_016868810.1
43
5723
XM_017013322.1
XP_016868811.1
42
5685
XM_017013323.1
XP_016868812.1
43
5699
XM_017013324.1
XP_016868813.1
41
5812
XM_017013325.1
XP_016868814.1
42
5624
XM_017013326.1
XP_016868815.1
41
5572
XM_017013327.1
XP_016868816.1
43
5292
XM_017013328.1
XP_016868817.1
42
5247
XM_017013329.1
XP_016868818.1
41
5324
GenBank mRNA
AB209418.1
-
41
5835
AK126647.1
-
24
2908
AK129864.1
-
2
471
AK223578.1
-
2
439
BC007930.2
-
2
493
BC030957.1
-
2
506
BC117121.1
-
2
281
BX647936.1
-
18
2827
M28880.1
-
40
5514
X16609.1
-
40
5514
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
exocytosis
ER to Golgi vesicle-mediated transport
cytoskeleton organization
signal transduction
positive regulation of organelle organization
maintenance of epithelial cell apical/basal polarity
protein targeting to plasma membrane
structural molecule activity
structural constituent of cytoskeleton
protein binding
cytoskeletal adaptor activity
enzyme binding
spectrin binding
ATPase binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0