Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: TFPT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321792.1 | NP_001308721.1 | 4 | 407 | ||
NM_013342.3 | NP_037474.1 | 4 | 801 | |||
XM_005278261.1 | XP_005278318.1 | 3 | 277 | |||
GenBank mRNA | AF052052.1 | - | 5 | 674 | ||
BC004281.1 | - | 4 | 433 | 3616596 | ||
BC007776.2 | - | 4 | 419 | 4301790 | ||
BQ027375.1 | - | 3 | 419 |