Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other GUK1 SIRNA Assays ›
          Gene Symbol:
          GUK1
          Gene Name:
          guanylate kinase 1
          Gene Aliases:
          GMK
          Chromosome Location:
          Chr.1: 228139643 - 228148955 on Build GRCh38
          UniGene ID:
          Hs.376933
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS179080
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price 459.00
          Your Price
          Online offer:
          459.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: GUK1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000858.5 NP_000849.1 6 620
          NM_001159390.1 NP_001152862.1 5 455
          NM_001159391.1 NP_001152863.1 5 369
          NM_001242839.1 NP_001229768.1 5 395
          NM_001242840.1 NP_001229769.1 5 455
          XM_005273103.3 XP_005273160.1 7 791
          XM_005273104.4 XP_005273161.1 7 789
          XM_005273105.3 XP_005273162.1 6 648
          XM_005273106.2 XP_005273163.1 5 836
          XM_011544174.2 XP_011542476.1 5 3219
          XM_017001091.1 XP_016856580.1 7 773
          XM_017001092.1 XP_016856581.1 6 1108
          XM_017001093.1 XP_016856582.1 6 568
          XM_017001094.1 XP_016856583.1 5 406
          XM_017001095.1 XP_016856584.1 6 565
          XM_017001096.1 XP_016856585.1 5 405
          GenBank mRNA AK303845.1 - 5 408
          AL832986.1 - 2 2131
          BC006249.2 - 6 558 3946293
          BC007369.2 - 4 272
          BC009914.2 - 6 532 2821563
          BC091480.1 - 5 375 30340102
          BE780558.1 - 5 342
          BI771824.1 - 5 395
          BU194785.1 - 421
          CR456836.1 - 4 317
          CR542006.1 - 4 317
          L76200.1 - 4 317
          U66895.1 - 4 323

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          purine nucleotide metabolic process
          dGDP biosynthetic process
          nucleobase-containing small molecule interconversion
          GDP-mannose metabolic process
          glycoprotein transport
          ATP metabolic process
          GMP metabolic process
          dGMP metabolic process
          dATP metabolic process
          GDP biosynthetic process
          nucleotide phosphorylation
          guanylate kinase activity
          ATP binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0