Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: TIMM22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013337.2 | NP_037469.2 | 3 | 482 | ||
GenBank mRNA | AK000830.1 | - | 3 | 463 | ||
AK225453.1 | - | 3 | 463 | |||
BC002324.2 | - | 3 | 482 | 2819655 |