Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other HNRNPC SIRNA Assays ›
          Gene Symbol:
          HNRNPC
          Gene Name:
          heterogeneous nuclear ribonucleoprotein C (C1/C2)
          Gene Aliases:
          C1, C2, HNRNP, HNRPC, SNRPC
          Chromosome Location:
          Chr.14: 21209136 - 21269479 on Build GRCh38
          UniGene ID:
          Hs.508848
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS179305
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price Contact Us
          Your Price
          Online offer:
          3062.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: HNRNPC
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001077442.1 NP_001070910.1 6, 7 899
          NM_001077443.1 NP_001070911.1 6, 7 860
          NM_004500.3 NP_004491.2 7, 8 886
          NM_031314.2 NP_112604.2 7, 8 925
          XM_006720125.2 XP_006720188.1 7, 8 1542
          XM_011536708.1 XP_011535010.1 8, 9 959
          XM_011536709.2 XP_011535011.1 7, 8 1380
          XM_011536710.2 XP_011535012.1 7, 8 1581
          XM_011536711.2 XP_011535013.1 7, 8 1588
          XM_011536712.1 XP_011535014.1 8, 9 920
          XM_017021252.1 XP_016876741.1 7, 8 1549
          XM_017021253.1 XP_016876742.1 7, 8 1340
          GenBank mRNA AB209527.1 - 7, 8 859
          AK126950.1 - 6, 7 3594
          AK223517.1 - 7, 8 864
          AK225032.1 - 7, 8 849
          AK292619.1 - 9, 10 1202
          AK297482.1 - 5, 6 725
          AK298906.1 - 6, 7 557
          AK299923.1 - 6, 7 749
          AK302213.1 - 6, 7 823
          AK312338.1 - 7, 8 831
          AK312382.1 - 6, 7 845
          AL833432.1 - 4, 5 462
          BC003394.1 - 7, 8 817 3447724
          BC007052.1 - 6, 7 845 3686841
          BC008423.1 - 6, 7 845 4102910
          BC066932.1 - 6, 7 805 4375450
          BC089438.1 - 6, 7 652 30555817
          BC103758.1 - 6, 7 805 6187512
          BC108658.1 - 6, 7 750 6501821
          BX161480.1 - 6, 7 755
          BX247961.1 - 6, 7 616
          BX247992.1 - 6, 7 771
          M16342.1 - 7, 8 764
          M29063.1 - 7, 8 860

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          mRNA splicing, via spliceosome
          osteoblast differentiation
          RNA splicing
          gene expression
          protein sumoylation
          negative regulation of telomere maintenance via telomerase
          ATP-dependent chromatin remodeling
          3'-UTR-mediated mRNA stabilization
          nucleotide binding
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          RNA polymerase II distal enhancer sequence-specific DNA binding
          RNA binding
          mRNA 3'-UTR binding
          protein binding
          poly(U) RNA binding
          nucleosomal DNA binding
          identical protein binding
          poly(A) RNA binding
          telomerase RNA binding
          N6-methyladenosine-containing RNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0