Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLHL38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081675.2 | NP_001075144.2 | 1 | 41 | ||
XM_005250901.4 | XP_005250958.1 | 2 | 262 | |||
GenBank mRNA | AB196636.1 | - | 1 | 17 | ||
BC127886.1 | - | 1 | 41 | 40131758 |