This product has been added to your favorites list. Go to
My Favorites
LOADING 
- Gene Symbol:
-
GLDN
- Gene Name:
-
gliomedin
- Gene Aliases:
-
CLOM, COLM, CRG-L2, CRGL2, UNC-112
- Chromosome Location:
-
Chr.15: 51341516 - 51413365 on Build GRCh38
- UniGene ID:
-
Hs.526441
- Species:
- Human
- Species Specific ID (Flybase ID):
-
-
siRNA Details
|
Gene Symbol:
GLDN |
| Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
| RefSeq |
NM_181789.2 |
NP_861454.2 |
6, 7
|
994
|
|
| XM_011521501.2 |
XP_011519803.1 |
6, 7
|
2851
|
|
| XM_017022121.1 |
XP_016877610.1 |
5, 6
|
837
|
|
| XM_017022122.1 |
XP_016877611.1 |
6, 7
|
1025
|
|
| XM_017022123.1 |
XP_016877612.1 |
6, 7
|
816
|
|
| XM_017022124.1 |
XP_016877613.1 |
6, 7
|
868
|
|
| XM_017022125.1 |
XP_016877614.1 |
6, 7
|
855
|
|
| XM_017022126.1 |
XP_016877615.1 |
6, 7
|
868
|
|
| GenBank mRNA |
AK172756.1 |
- |
6, 7
|
830
|
|
| AY358144.1 |
- |
5, 6
|
455
|
|
| BC113397.1 |
- |
6, 7
|
480
|
8322449 |
| BC113399.1 |
- |
6, 7
|
480
|
8322451 |
| BX538105.1 |
- |
6, 7
|
503
|
|
Back To Top
Related Products
-
Anti-GAPDH, Mouse Monoclonal 6C5
-
Anti-β-Actin, Mouse Monoclonal AC-15
-
TaqMan® Gene Expression Cells-to-CT™
-
Neon™ Transfection System 100 µL Kit
-
Lipofectamine™ RNAiMAX Transfection Reagent
-
Invivofectamine 2.0
-
Lipofectamine™ 2000 Transfection Reagent
-
Neon™ Transfection System