Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other IL15RA SIRNA Assays ›
Gene Symbol:
IL15RA
Gene Name:
interleukin 15 receptor subunit alpha
Gene Aliases:
CD215
Chromosome Location:
Chr.10: 5948897 - 5978741 on Build GRCh38
UniGene ID:
Hs.445124
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS179911
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: IL15RA
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001243539.1
NP_001230468.1
4
747
NM_001256765.1
NP_001243694.1
5
696
NM_002189.3
NP_002180.1
4
543
NM_172200.2
NP_751950.2
3
444
NR_046362.1
2
453
XM_011519461.2
XP_011517763.1
6
937
XM_011519462.2
XP_011517764.1
5
855
XM_011519463.2
XP_011517765.1
5
838
XM_011519464.2
XP_011517766.1
5
784
XM_011519465.1
XP_011517767.1
6
937
XM_011519466.2
XP_011517768.1
6
937
XM_011519467.2
XP_011517769.1
4
700
XM_011519468.1
XP_011517770.1
6
937
XM_011519469.1
XP_011517771.1
5
838
XM_011519470.2
XP_011517772.1
3
603
XM_011519471.2
XP_011517773.1
4
668
XM_011519472.2
XP_011517774.1
3
507
XM_011519474.2
XP_011517776.1
3
490
XM_011519475.2
XP_011517777.1
2
408
XM_011519476.2
XP_011517778.1
2
398
XM_011519477.2
XP_011517779.1
2
403
XM_017016195.1
XP_016871684.1
5
784
XM_017016196.1
XP_016871685.1
4
509
XM_017016197.1
XP_016871686.1
5
524
XM_017016198.1
XP_016871687.1
4
668
XM_017016200.1
XP_016871689.1
3
1095
XM_017016201.1
XP_016871690.1
4
792
XM_017016202.1
XP_016871691.1
5
513
XM_017016203.1
XP_016871692.1
3
1095
GenBank mRNA
AF035279.1
-
4
478
AK304211.1
-
3
355
BC065506.1
-
2
445
BC074726.2
-
4
455
30915179
BC107777.1
-
4
611
6497994
BC121140.2
-
4
450
40120998
BC121141.2
-
4
455
40120999
BM546106.1
-
2
464
CR457064.1
-
3
316
CR542023.1
-
4
424
DC298692.1
-
3
449
HQ401283.1
-
5
577
JX987304.1
-
5
577
JX987305.1
-
5
577
JX987306.1
-
5
577
JX987307.1
-
5
577
JX987308.1
-
5
577
JX987309.1
-
5
577
JX987310.1
-
5
577
JX987311.1
-
5
577
U31628.1
-
4
506
Back To Top
More Information
Panther Classification:
Molecular Function -
transmembrane signal receptor
Gene Ontology Categories:
Function(s)
Process(es)
signal transduction
cell proliferation
cytokine-mediated signaling pathway
signal transducer activity
cytokine receptor activity
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-jmx4f:80/100.66.76.53:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0