Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other LIG4 SIRNA Assays ›
          Gene Symbol:
          LIG4
          Gene Name:
          DNA ligase 4
          Gene Aliases:
          LIG4S
          Chromosome Location:
          Chr.13: 108207442 - 108218368 on Build GRCh38
          UniGene ID:
          Hs.166091
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS180681
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: LIG4
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001098268.1 NP_001091738.1 2 2690
          NM_002312.3 NP_002303.2 2 2728
          NM_206937.1 NP_996820.1 3 2607
          XM_005254056.1 XP_005254113.1 3 2572
          XM_005254057.4 XP_005254114.1 3 2742
          XM_005254058.3 XP_005254115.1 2 4188
          XM_006719951.3 XP_006720014.1 3 4447
          XM_006719952.1 XP_006720015.1 4 3040
          XM_011521091.2 XP_011519393.1 4 3069
          XM_011521092.2 XP_011519394.1 3 2645
          XM_017020563.1 XP_016876052.1 3 4289
          XM_017020564.1 XP_016876053.1 2 2715
          XM_017020565.1 XP_016876054.1 4 3086
          XM_017020566.1 XP_016876055.1 4 2989
          XM_017020567.1 XP_016876056.1 3 2643
          XM_017020568.1 XP_016876057.1 3 4265
          XM_017020569.1 XP_016876058.1 4 4710
          XM_017020570.1 XP_016876059.1 4 2989
          XM_017020571.1 XP_016876060.1 4 4794
          XM_017020572.1 XP_016876061.1 3 4181
          XM_017020573.1 XP_016876062.1 2 2491
          GenBank mRNA AK292419.1 - 3 2516
          AK314844.1 - 3 2572
          BC037491.2 - 3 2607 5259632
          BX648833.1 - 2 2690
          X83441.1 - 2 2728

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          DNA metabolism protein

          Gene Ontology Categories:

          Function(s) Process(es)

          single strand break repair
          in utero embryonic development
          pro-B cell differentiation
          DNA ligation
          lagging strand elongation
          nucleotide-excision repair, DNA gap filling
          double-strand break repair
          double-strand break repair via nonhomologous end joining
          cell cycle
          central nervous system development
          cell proliferation
          response to X-ray
          response to gamma radiation
          T cell differentiation in thymus
          V(D)J recombination
          immunoglobulin V(D)J recombination
          T cell receptor V(D)J recombination
          somatic stem cell population maintenance
          negative regulation of neuron apoptotic process
          isotype switching
          positive regulation of fibroblast proliferation
          positive regulation of neurogenesis
          DNA ligation involved in DNA recombination
          DNA ligation involved in DNA repair
          chromosome organization
          cell division
          neuron apoptotic process
          cellular response to lithium ion
          DNA biosynthetic process
          establishment of integrated proviral latency
          double-strand break repair via classical nonhomologous end joining
          positive regulation of chromosome organization
          DNA binding
          DNA ligase activity
          DNA ligase (ATP) activity
          protein binding
          ATP binding
          protein C-terminus binding
          ligase activity
          metal ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0