Search Thermo Fisher Scientific
Gene Symbol: LMO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270428.1 | NP_001257357.1 | 4 | 613 | ||
NM_002315.2 | NP_002306.1 | 4 | 945 | |||
NR_073006.1 | 4 | 1066 | ||||
XM_006718228.3 | XP_006718291.1 | 4 | 1991 | |||
XM_011520098.2 | XP_011518400.1 | 4 | 480 | |||
XM_011520099.2 | XP_011518401.1 | 4 | 497 | |||
XM_011520100.1 | XP_011518402.1 | 3 | 344 | |||
GenBank mRNA | AW006226.1 | - | 1 | 367 | ||
BC039512.1 | - | 4 | 613 | |||
BC069673.1 | - | 4 | 487 | 7262648 | ||
BC069752.1 | - | 4 | 821 | 7262660 | ||
BC069793.1 | - | 4 | 821 | 7262577 | ||
BC096056.3 | - | 4 | 455 | 40000075 | ||
BC096057.3 | - | 4 | 454 | 40000076 | ||
BC096058.1 | - | 4 | 576 | 40000078 | ||
M26682.1 | - | 4 | 926 | |||
X64244.1 | - | 4 | 429 | |||
X64247.1 | - | 1 | 80 |