Search
Search

SMAD3| Gene Symbol: SMAD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145102.1 | NP_001138574.1 | 4 | 639 | ||
| NM_001145103.1 | NP_001138575.1 | 4 | 450 | |||
| NM_001145104.1 | NP_001138576.1 | 2 | 83 | |||
| NM_005902.3 | NP_005893.1 | 4 | 874 | |||
| XM_011521559.2 | XP_011519861.1 | 3 | 901 | |||
| XM_011521560.1 | XP_011519862.1 | 4 | 813 | |||
| GenBank mRNA | AB209616.1 | - | 4 | 459 | ||
| AK290881.1 | - | 5 | 799 | |||
| AK298139.1 | - | 4 | 450 | |||
| AK300614.1 | - | 2 | 80 | |||
| AK301348.1 | - | 2 | 75 | |||
| AK316017.1 | - | 4 | 499 | |||
| BC050743.1 | - | 4 | 792 | 6094328 | ||
| DA977882.1 | - | 2 | 83 | |||
| EU016553.1 | - | 2 | 86 | |||
| U68019.1 | - | 4 | 642 | |||
| U76622.1 | - | 4 | 613 | |||