Search
Search

MAGEA1| Gene Symbol: MAGEA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004988.4 | NP_004979.3 | 3 | 496 | ||
| GenBank mRNA | AK314979.1 | - | 3 | 496 | ||
| AY148486.1 | - | 3 | 454 | |||
| BC017555.1 | - | 3 | 463 | 3922845 | ||
| BP314195.1 | - | 2 | 417 | |||
| BT009789.1 | - | 1 | 276 | |||
| DB053667.1 | - | 3 | 510 | |||