Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Instrument Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
Our Instagram
Our Facebook
Blog Behind the Bench
Customer Experience Center (CEC)
Ecommerce Exclusives
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MAP2 SIRNA Assays ›
Gene Symbol:
MAP2
Gene Name:
microtubule associated protein 2
Gene Aliases:
MAP2A, MAP2B, MAP2C
Chromosome Location:
Chr.2: 209424047 - 209734118 on Build GRCh38
UniGene ID:
Hs.368281
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS181001
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MAP2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001039538.1
NP_001034627.1
5
639
NM_002374.3
NP_002365.3
4
436
NM_031845.2
NP_114033.2
4
436
NM_031847.2
NP_114035.2
4
436
XM_005246555.1
XP_005246612.1
4
555
XM_006712532.1
XP_006712595.1
4
555
XM_011511186.1
XP_011509488.1
4
462
XM_011511187.1
XP_011509489.1
3
359
XM_011511188.1
XP_011509490.1
3
359
XM_011511190.1
XP_011509492.1
3
359
XM_011511191.1
XP_011509493.1
3
359
XM_011511192.1
XP_011509494.1
3
359
XM_011511193.1
XP_011509495.1
3
359
XM_011511195.1
XP_011509497.1
3
359
XM_011511196.2
XP_011509498.1
4
555
XM_011511197.2
XP_011509499.1
4
555
XM_017004111.1
XP_016859600.1
3
359
XM_017004112.1
XP_016859601.1
4
555
XM_017004113.1
XP_016859602.1
4
462
XM_017004114.1
XP_016859603.1
4
555
XM_017004115.1
XP_016859604.1
3
359
XM_017004116.1
XP_016859605.1
3
359
XM_017004117.1
XP_016859606.1
3
359
XM_017004118.1
XP_016859607.1
4
462
XM_017004119.1
XP_016859608.1
4
555
XM_017004120.1
XP_016859609.1
4
555
XM_017004121.1
XP_016859610.1
4
462
XM_017004122.1
XP_016859611.1
4
462
XM_017004123.1
XP_016859612.1
4
555
XM_017004124.1
XP_016859613.1
4
462
XM_017004125.1
XP_016859614.1
4
555
XM_017004126.1
XP_016859615.1
4
555
XM_017004127.1
XP_016859616.1
4
555
XM_017004128.1
XP_016859617.1
3
359
XM_017004129.1
XP_016859618.1
3
359
XM_017004138.1
XP_016859627.1
4
462
XM_017004139.1
XP_016859628.1
1
219
XM_017004140.1
XP_016859629.1
4
462
GenBank mRNA
AB209330.1
-
3
354
AK291446.1
-
1
225
AK307603.1
-
4
414
BC027583.1
-
3
339
BC038857.1
-
5
628
BC066648.1
-
5
577
BC110423.1
-
4
684
BC117123.1
-
2
265
BC143245.1
-
2
270
U01828.1
-
4
436
U89329.1
-
1
188
U89330.1
-
1
188
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
microtubule cytoskeleton organization
microtubule bundle formation
central nervous system neuron development
neuron projection development
dendrite morphogenesis
dystroglycan binding
structural molecule activity
protein binding
calmodulin binding
microtubule binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-2dmq6:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0