Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MARK3 SIRNA Assays ›
Gene Symbol:
MARK3
Gene Name:
microtubule affinity regulating kinase 3
Gene Aliases:
CTAK1, KP78, PAR1A, Par-1a
Chromosome Location:
Chr.14: 103385364 - 103503831 on Build GRCh38
UniGene ID:
Hs.35828
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS181007
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MARK3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001128918.2
NP_001122390.2
5
1063
NM_001128919.2
NP_001122391.2
5
1063
NM_001128920.2
NP_001122392.2
5
1063
NM_001128921.2
NP_001122393.2
5
1063
NM_002376.6
NP_002367.5
5
1063
XM_005267641.4
XP_005267698.2
5
442
XM_005267642.4
XP_005267699.2
5
1063
XM_005267643.4
XP_005267700.2
5
1063
XM_006720146.3
XP_006720209.1
5
437
XM_011536769.2
XP_011535071.1
4
176
XM_017021291.1
XP_016876780.1
5
1063
XM_017021292.1
XP_016876781.1
5
1063
XM_017021293.1
XP_016876782.1
6
538
XM_017021294.1
XP_016876783.1
4
812
XM_017021295.1
XP_016876784.1
4
827
XM_017021296.1
XP_016876785.1
4
844
XM_017021297.1
XP_016876786.1
4
820
XM_017021298.1
XP_016876787.1
4
820
XM_017021299.1
XP_016876788.1
4
871
XM_017021300.1
XP_016876789.1
3
297
XM_017021301.1
XP_016876790.1
3
452
XM_017021302.1
XP_016876791.1
3
297
XM_017021303.1
XP_016876792.1
3
297
XM_017021304.1
XP_016876793.1
3
297
XM_017021305.1
XP_016876794.1
3
297
XM_017021306.1
XP_016876795.1
5
1063
XM_017021307.1
XP_016876796.1
4
1014
XM_017021308.1
XP_016876797.1
4
1014
XM_017021309.1
XP_016876798.1
4
346
XM_017021310.1
XP_016876799.1
4
346
XM_017021311.1
XP_016876800.1
3
297
GenBank mRNA
AF387637.1
-
5
415
AF465413.1
-
5
907
AK307448.1
-
5
442
BC024773.1
-
5
886
5138958
BX161395.1
-
5
993
M80359.1
-
5
568
U64205.1
-
5
772
Back To Top
More Information
Panther Classification:
Molecular Function -
non-receptor serine/threonine protein kinase
Gene Ontology Categories:
Function(s)
Process(es)
MAPK cascade
microtubule cytoskeleton organization
establishment of cell polarity
protein serine/threonine kinase activity
protein binding
ATP binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-fz59j:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0