Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: TSPAN11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080509.2 | NP_001073978.1 | 5, 6 | 565 | ||
XM_011520679.2 | XP_011518981.1 | 5, 6 | 827 | |||
XM_011520680.2 | XP_011518982.1 | 5, 6 | 805 | |||
GenBank mRNA | AK293947.1 | - | 5, 6 | 512 | ||
AK300920.1 | - | 4, 5 | 470 | |||
BC127660.1 | - | 3, 4 | 331 | |||
BC127661.1 | - | 3, 4 | 331 | |||
BC146917.1 | - | 5, 6 | 511 | 9056845 | ||
BC146922.1 | - | 5, 6 | 511 | 9056850 |