Search
Search

TARM1| Gene Symbol: TARM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135686.2 | NP_001129158.2 | 4 | 655 | ||
| XM_005278278.3 | XP_005278335.1 | 4 | 924 | |||
| GenBank mRNA | AK301730.1 | - | 4 | 940 | ||
| DQ479398.1 | - | 4 | 655 | |||