Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Enterprise Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other NBL1 SIRNA Assays ›
          Gene Symbol:
          NBL1 MINOS1-NBL1
          Gene Name:
          neuroblastoma 1, DAN family BMP antagonist
          MINOS1-NBL1 readthrough
          Gene Aliases:
          D1S1733E, DAN, DAND1, NB, NO3
          C1orf151-NBL1
          Chromosome Location:
          Chr.1: 19643229 - 19658456 on Build GRCh38
          Chromosome Location:
          Chr.1: 19596977 - 19658456 on Build GRCh38
          UniGene ID:
          Hs.654502
          Hs.466662
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID HSS181439
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: NBL1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001204084.2 NP_001191013.1 2 355
          NM_001204085.1 NP_001191014.1 2 273
          NM_001204086.2 NP_001191015.1 2 382
          NM_001278164.1 NP_001265093.1 2 264
          NM_001278165.1 NP_001265094.1 2 331
          NM_001278166.1 NP_001265095.1 2 175
          NM_005380.7 NP_005371.2 2 388
          NM_182744.3 NP_877421.2 2 283
          GenBank mRNA AK292101.1 - 2 314
          AK300872.1 - 2 283
          AK309257.1 - 2 382
          AK313265.1 - 2 273
          AL703187.1 - 2 281
          AY049783.1 - 1 112
          BC012037.1 - 2 184 3874204
          BQ671197.1 - 240
          BT019423.1 - 1 92
          D28124.1 - 2 153
          DA384331.1 - 2 331
          DA921297.1 - 2 280
          DA924824.1 - 2 355
          DA929341.1 - 2 179
          DB016315.1 - 2 262
          DB253522.1 - 2 283
          DB482000.1 - 2 248
          HY084536.1 - 2 171
          Gene Symbol: MINOS1-NBL1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001204088.1 NP_001191017.1 3 432
          NM_001204089.1 NP_001191018.1 2 247
          GenBank mRNA AK289456.1 - 2 245
          FY212253.1 - 3 341

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          nervous system development
          negative regulation of BMP signaling pathway
          sequestering of BMP in extracellular matrix
          sequestering of BMP from receptor via BMP binding
          positive regulation of neuron differentiation
          determination of dorsal identity
          neuron projection morphogenesis
          negative regulation of monocyte chemotaxis
          protein binding
          morphogen activity
          BMP binding
          protein homodimerization activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-mtn5c:80/100.66.73.196:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0