Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: YBX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015982.3 | NP_057066.2 | 4, 5 | 524 | ||
XM_017024713.1 | XP_016880202.1 | 5, 6 | 614 | |||
GenBank mRNA | AB208818.1 | - | 1, 2 | 3505 | ||
AF096834.1 | - | 4, 5 | 522 | |||
AK092747.1 | - | 3, 4 | 1141 | |||
AK315035.1 | - | 4, 5 | 495 | |||
BC033800.1 | - | 4, 5 | 487 | 5171696 | ||
BC047760.1 | - | 4, 5 | 488 | 5742121 |